2-Brom-N,N-dimethyl-5-fluorbenzamid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-fluoro-N,N-dimethylbenzamide

PC8019-1G PC8019-5G PC8019-250MG
APOSPC8019-1GEA 190 EUR
APOSPC8019-1G APOSPC8019-5G APOSPC8019-250MG
2-Brom-N,N-dimethyl-5-fluorbenzamid 98%
2-Brom-N,N-dimethyl-5-fluorbenzamid
Formel: C₉H₉BrFNO
Molecular Weight: 246.0763 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800904
CAS-Nummer: 951884-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR