4-(Trifluormethyl)benzoesäurehydrazid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethyl)benzoylhydrazine, 4-(Trifluoromethyl)benzhydrazide, 4-(Trifluoromethyl)benzoic Hydrazide, 4-(Trifluoromethyl)benzohydrazide
PC8021-25G
PC8021-5G
APOSPC8021-25GEA
102
EUR
APOSPC8021-25G
APOSPC8021-5G
4-(Trifluormethyl)benzoesäurehydrazid 95%
4-(Trifluoromethyl)benzoic acid hydrazide
Formel:
C₈H₇F₃N₂O Schmelzpunkt: 115…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051703 CAS-Nummer: 339-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...