4-(2-Bromo-5-fluorobenzoyl)morpholine
Lieferant: Apollo Scientific
PC8027-1G
APOSPC8027-1GEA
176
EUR
APOSPC8027-1G
4-(2-Bromo-5-fluorobenzoyl)morpholine
4-(2-Bromo-5-fluorobenzoyl)morpholine
Formel:
C₁₁H₁₁BrFNO₂ Molecular Weight: 288,113 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800908 CAS-Nummer: 951884-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...