2-Brom-5-fluor-N-phenylbenzamid
Lieferant: Apollo Scientific
PC8029-1G
APOSPC8029-1GEA
176
EUR
APOSPC8029-1G
2-Brom-5-fluor-N-phenylbenzamid
2-Brom-5-fluor-N-phenylbenzamid
Formel:
C₁₃H₉BrFNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800907 CAS-Nummer: 949443-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...