4-Carbamoyl-2-fluorphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Carbamoyl-2-fluorophenyl)boronic acid

PC8031-1G
APOSPC8031-1GEA 319 EUR
APOSPC8031-1G
4-Carbamoyl-2-fluorphenylboronsäure
4-Carbamoyl-2-fluorphenylboronsäure
Formel: C₇H₇BFNO₃
Molecular Weight: 182,9500 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09800867
CAS-Nummer: 874289-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR