Zinc hexafluoro-2,4-pentanedionate dihydrate 98%

Lieferant: Apollo Scientific

Synonyme: Zinc hexafluoroacetylacetonate hydrate

PC8032-5G
APOSPC8032-5GEA 350 EUR
APOSPC8032-5G
Zinc hexafluoro-2,4-pentanedionate dihydrate 98%
Zinc hexafluoro-2,4-pentanedionate dihydrate
Formel: C₁₀H₂F₁₂O₄Zn.2H₂O
Molecular Weight: 515,541 g/mol
Schmelzpunkt: 157…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150577
CAS-Nummer: 16743-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR