2-Brom-5-fluor-N-propylbenzamid

Lieferant: Apollo Scientific

PC8033-1G
APOSPC8033-1GEA 176 EUR
APOSPC8033-1G
2-Brom-5-fluor-N-propylbenzamid
2-Brom-5-fluor-N-propylbenzamid
Formel: C₁₀H₁₁BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09800906
CAS-Nummer: 951884-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR