1-(2-Bromo-5-fluorobenzoyl)pyrrolidine

Lieferant: Apollo Scientific

Synonyme: (2-Bromo-5-fluorophenyl)(pyrrolidin-1-yl)methanone

PC8036-1G
APOSPC8036-1GEA 176 EUR
APOSPC8036-1G
1-(2-Bromo-5-fluorobenzoyl)pyrrolidine
1-(2-Bromo-5-fluorobenzoyl)pyrrolidine
Formel: C₁₁H₁₁BrFNO
Molecular Weight: 272,114 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800909
CAS-Nummer: 951884-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR