5-Bromo-2-fluoro-3-methylbenzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: (5-Bromo-2-fluoro-3-methylphenyl)boronic acid, 5-Bromo-2-fluoro-3-methylphenylboronic acid

PC8046-1G PC8046-5G
APOSPC8046-1GEA 63.8 EUR
APOSPC8046-1G APOSPC8046-5G
5-Bromo-2-fluoro-3-methylbenzeneboronic acid
5-Bromo-2-fluoro-3-methylbenzeneboronic acid
Formel: C₇H₇BBrFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800868
CAS-Nummer: 957120-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR