1-(2-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}ethyl)piperazine

Lieferant: Apollo Scientific
Achtung

PC8060-1G
APOSPC8060-1GEA 277 EUR
APOSPC8060-1G
1-(2-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}ethyl)piperazine
1-(2-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}ethyl)piperazine
Formel: C₁₂H₁₆F₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09836181
CAS-Nummer: 1000339-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR