2-(Aminomethyl)-4'-fluorobiphenyl 95%

Lieferant: Apollo Scientific

Synonyme: (4'-Fluoro-[1,1'-biphenyl]-2-yl)methanamine hydrochloride, 1-(4'-Fluorobiphenyl-2-yl)methylamine hydrochloride

PC8062-250MG
APOSPC8062-250MGEA 106 EUR
APOSPC8062-250MG
2-(Aminomethyl)-4'-fluorobiphenyl 95%
2-(Aminomethyl)-4'-fluorobiphenyl
Formel: C₁₃H₁₂FN·ClH
Molecular Weight: 237,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02089421
CAS-Nummer: 1189729-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR