2-Brom-3-(trifluormethyl)anilin 98%

Lieferant: Apollo Scientific
Achtung

PC8075-5G PC8075-1G PC8075-25G
APOSPC8075-5GEA 29 EUR
APOSPC8075-5G APOSPC8075-1G APOSPC8075-25G
2-Brom-3-(trifluormethyl)anilin 98%
2-Brom-3-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
Molecular Weight: 240.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01656033
CAS-Nummer: 58458-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR