2-Amino-6-fluorbenzothiazol 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC8076-25G
PC8076-5G
PC8076-1G
PC8076-100G
APOSPC8076-25GEA
94.4
EUR
APOSPC8076-25G
APOSPC8076-5G
APOSPC8076-1G
APOSPC8076-100G
2-Amino-6-fluorbenzothiazol 99%
2-Amino-6-fluorobenzothiazole
Formel:
C₇H₅FN₂S Schmelzpunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013336 CAS-Nummer: 348-40-3 EINECS: 609-034-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...