N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid

Lieferant: Apollo Scientific

Synonyme: 3-(N-Benzylsulphamoyl)-5-bromobenzotrifluoride

PC8081-1G
APOSPC8081-1GEA 212 EUR
APOSPC8081-1G
N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid
N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid
Formel: C₁₄H₁₁BrF₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09800987
CAS-Nummer: 951885-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR