4-[N-(4-Fluoro-3-methoxyphenyl)sulphamoyl]benzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: (4-(N-(4-Fluoro-3-methoxyphenyl)sulphamoyl)phenyl)boronic acid

PC8084-1G
APOSPC8084-1GEA 452 EUR
APOSPC8084-1G
4-[N-(4-Fluoro-3-methoxyphenyl)sulphamoyl]benzeneboronic acid
4-[N-(4-Fluoro-3-methoxyphenyl)sulphamoyl]benzeneboronic acid
Formel: C₁₃H₁₃BFNO₅S
Molecular Weight: 325,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800884
CAS-Nummer: 957120-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR