5-Fluor-3-(4-hydroxybutyl)oxindol

Lieferant: Apollo Scientific

PC8090-50MG
APOSPC8090-50MGEA 314 EUR
APOSPC8090-50MG
5-Fluor-3-(4-hydroxybutyl)oxindol
5-Fluor-3-(4-hydroxybutyl)oxindol
Formel: C₁₂H₁₄FNO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 637341-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR