3-Brom-5-iodbenzotrifluorid 98%

Lieferant: Apollo Scientific
Achtung

PC8091-1G PC8091-5G PC8091-10G
APOSPC8091-1GEA 21.9 EUR
APOSPC8091-1G APOSPC8091-5G APOSPC8091-10G
3-Brom-5-iodbenzotrifluorid 98%
3-Brom-5-iodbenzotrifluorid
Formel: C₇H₃BrF₃I
Lagertemperatur: Kühlschrank
MDL: MFCD09033749
CAS-Nummer: 481075-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR