2,4-Dichloro-6-(3-fluorophenoxy)-1,3,5-triazine

Lieferant: Apollo Scientific
Achtung

PC8098-5G
APOSPC8098-5GEA 313 EUR
APOSPC8098-5G
2,4-Dichloro-6-(3-fluorophenoxy)-1,3,5-triazine
2,4-Dichloro-6-(3-fluorophenoxy)-1,3,5-triazine
Formel: C₉H₄Cl₂FN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD05154988
CAS-Nummer: 112748-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR