4-(3-Brom-5-(trifluormethyl)phenylsulfonyl)morpholin

Lieferant: Apollo Scientific

PC8103-1G
APOSPC8103-1GEA 311 EUR
APOSPC8103-1G
4-(3-Brom-5-(trifluormethyl)phenylsulfonyl)morpholin
4-(3-Brom-5-(trifluormethyl)phenylsulfonyl)morpholin
Formel: C₁₁H₁₁BrF₃NO₃S
Schmelzpunkt: 131…133 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09801007
CAS-Nummer: 951884-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR