3-Brom-N-(furan-2-ylmethyl)-5-(trifluormethyl)benzolsulfonamid

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-5-[N-(fur-2-ylmethyl)sulphamoyl]benzotrifluoride

PC8105-1G
APOSPC8105-1GEA 248 EUR
APOSPC8105-1G
3-Brom-N-(furan-2-ylmethyl)-5-(trifluormethyl)benzolsulfonamid
3-Brom-N-(furan-2-ylmethyl)-5-(trifluormethyl)benzolsulfonamid
Formel: C₁₂H₉BrF₃NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD09801006
CAS-Nummer: 951884-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR