3-Chlor-4-nitrobenzotrifluorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chlor-1-nitro-4-(trifluormethyl)benzol, 3-Chlor-4-nitrobenzotrifluorid, 1-Chlor-2-nitro-5-(trifluormethyl)benzol
PC8117-1G
PC8117-25G
PC8117-5G
APOSPC8117-1GEA
21.9
EUR
APOSPC8117-1G
APOSPC8117-25G
APOSPC8117-5G
3-Chlor-4-nitrobenzotrifluorid 97%
3-Chlor-4-nitrobenzotrifluorid
Formel:
C₇H₃ClF₃NO₂ Siedepunkt: 54 °C/ 0,4mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00272564 CAS-Nummer: 402-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...