4-Fluor-2-(trifluormethyl)zimtsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8123-1G
PC8123-10G
PC8123-5G
APOSPC8123-1GEA
29
EUR
APOSPC8123-1G
APOSPC8123-10G
APOSPC8123-5G
4-Fluor-2-(trifluormethyl)zimtsäure 97%
4-Fluor-2-(trifluormethyl)zimtsäure
Formel:
C₁₀H₆F₄O₂ Schmelzpunkt: 181…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236297 CAS-Nummer: 243977-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...