1-(4-Chlorobenzylamino)-2-[3-chloro-5-(trifluoromethyl)pyrid-2-ylamino]ethane 97%

Lieferant: Apollo Scientific

PC8128-500MG
APOSPC8128-500MGEA 688 EUR
APOSPC8128-500MG
1-(4-Chlorobenzylamino)-2-[3-chloro-5-(trifluoromethyl)pyrid-2-ylamino]ethane 97%
1-(4-Chlorobenzylamino)-2-[3-chloro-5-(trifluoromethyl)pyrid-2-ylamino]ethane
Formel: C₁₅H₁₄Cl₂F₃N₃
Molecular Weight: 364.193 g/mol
Schmelzpunkt: 41…43 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172583
CAS-Nummer: 338406-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR