5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde 97%

Lieferant: Apollo Scientific

PC8130-1G PC8130-500MG PC8130-5G
APOSPC8130-1GEA 533 EUR
APOSPC8130-1G APOSPC8130-500MG APOSPC8130-5G
5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde 97%
5-(2-Chlorobenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde
Formel: C₁₃H₁₀ClF₃N₂OS
Molecular Weight: 334.75 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00140828
CAS-Nummer: 321533-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR