Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate

PC8131-1G
APOSPC8131-1GEA 21.9 EUR
APOSPC8131-1G
Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Formel: C₁₃H₁₀F₃NO₃
Molecular Weight: 285,2186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878378
CAS-Nummer: 23851-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR