1-(2-Amino-6-chlorophenylamino)-2-[3-chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethane
Lieferant: Apollo Scientific
PC8135-500MG
APOSPC8135-500MGEA
688
EUR
APOSPC8135-500MG
1-(2-Amino-6-chlorophenylamino)-2-[3-chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethane
1-(2-Amino-6-chlorophenylamino)-2-[3-chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethane
Formel:
C₁₄H₁₃Cl₂F₃N₄ Molecular Weight: 365,181 g/mol Schmelzpunkt: 74…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793021 CAS-Nummer: 341966-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...