1-(2-Chloro-6-fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Chloro-6-fluorobenzyl)-5-cyano-6-methyl-1,2,3,4-tetrahydropyridin-2-one
PC8141-1G
APOSPC8141-1GEA
464
EUR
APOSPC8141-1G
1-(2-Chloro-6-fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile 97%
1-(2-Chloro-6-fluorobenzyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile
Formel:
C₁₄H₁₂ClFN₂O Schmelzpunkt: 109…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975042 CAS-Nummer: 338748-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...