2-Chlor-4-fluorzimtsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC8152-1G
PC8152-25G
PC8152-5G
APOSPC8152-1GEA
40.8
EUR
APOSPC8152-1G
APOSPC8152-25G
APOSPC8152-5G
2-Chlor-4-fluorzimtsäure 98%
2-Chlor-4-fluorzimtsäure
Formel:
C₉H₆ClFO₂ Schmelzpunkt: 247…249 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236027 CAS-Nummer: 133220-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...