1-Chlor-4-fluor-2-iodbenzol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8155-5G
APOSPC8155-5GEA
21.9
EUR
APOSPC8155-5G
1-Chlor-4-fluor-2-iodbenzol 97%
1-Chloro-4-fluoro-2-iodobenzene
Formel:
C₆H₃ClFI Molecular Weight: 256,4439 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00042575 CAS-Nummer: 202982-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...