4,5-Dibromo-2-fluorobenzoic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC8175-250MG
APOSPC8175-250MGEA
85.7
EUR
APOSPC8175-250MG
4,5-Dibromo-2-fluorobenzoic acid 95+%
4,5-Dibromo-2-fluorobenzoic acid
Formel:
C₇H₃Br₂FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672987 CAS-Nummer: 289039-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...