2,6-Dibrom-4-fluorphenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Dibromo-5-fluoro-2-hydroxybenzene

PC8177-5G
APOSPC8177-5GEA 21.9 EUR
APOSPC8177-5G
2,6-Dibrom-4-fluorphenol 97%
2,6-Dibrom-4-fluorphenol
Formel: C₆H₃Br₂FO
Molecular Weight: 269,9 g/mol
Schmelzpunkt: 54…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002317
CAS-Nummer: 344-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR