4,5-Dibrom-2-fluortoluol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Dibromo-4-fluoro-5-methylbenzene

PC8178-25G
APOSPC8178-25GEA 116 EUR
APOSPC8178-25G
4,5-Dibrom-2-fluortoluol
4,5-Dibrom-2-fluortoluol
Formel: C₇H₅Br₂F
Molecular Weight: 267,92 g/mol
Flammpunkt: 101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143234
CAS-Nummer: 202982-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR