4-Fluorbenzaldehydoxim

Lieferant: Apollo Scientific

PC8194-100G
APOSPC8194-100GEA 321 EUR
APOSPC8194-100G
4-Fluorbenzaldehydoxim
4-Fluorbenzaldehydoxim
Formel: C₇H₆FNO
Molecular Weight: 139,13 g/mol
Schmelzpunkt: 85…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00133101
CAS-Nummer: 459-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR