4-(Trifluormethoxy)benzotrifluorid 98%

Lieferant: Apollo Scientific
Achtung

PC8213-1G
APOSPC8213-1GEA 84.1 EUR
APOSPC8213-1G
4-(Trifluormethoxy)benzotrifluorid 98%
4-(Trifluormethoxy)benzotrifluorid
Formel: C₈H₄F₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00190125
CAS-Nummer: 80258-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR