2-(Trifluormethyl)anisol

Lieferant: Apollo Scientific
Achtung

PC8230-1G PC8230-250MG PC8230-5G
APOSPC8230-1GEA 87 EUR
APOSPC8230-1G APOSPC8230-250MG APOSPC8230-5G
2-(Trifluormethyl)anisol
2-(Trifluormethyl)anisol
Formel: C₈H₇F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00142938
CAS-Nummer: 395-48-2
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR