6-Fluor-4-chinolinol 98%

Lieferant: Apollo Scientific
Achtung

PC8235-1G PC8235-5G PC8235-10G
APOSPC8235-1GEA 24.8 EUR
APOSPC8235-1G APOSPC8235-5G APOSPC8235-10G
6-Fluor-4-chinolinol 98%
6-Fluor-4-chinolinol
Formel: C₉H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00269610
CAS-Nummer: 391-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR