6-Fluor-4-chinolinol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC8235-1G
PC8235-5G
PC8235-10G
APOSPC8235-1GEA
24.8
EUR
APOSPC8235-1G
APOSPC8235-5G
APOSPC8235-10G
6-Fluor-4-chinolinol 98%
6-Fluor-4-chinolinol
Formel:
C₉H₆FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269610 CAS-Nummer: 391-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...