4-Bromo-2,6-difluorophenylacetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Bromo-2,6-difluorophenyl)acetic acid

PC8245-5G
APOSPC8245-5GEA 148 EUR
APOSPC8245-5G
4-Bromo-2,6-difluorophenylacetic acid
4-Bromo-2,6-difluorophenylacetic acid
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251,027 g/mol
Schmelzpunkt: 150…152 °C
Dichte: 1,77 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD03094466
CAS-Nummer: 537033-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR