4-(Trifluoromethylthio)phenylacetic acid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-((Trifluoromethyl)thio)phenyl)acetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulphide, {4-[(Trifluoromethyl)sulphanyl]phenyl}acetic acid, 4-[(Trifluoromethyl)sulphanyl]phenylacetic acid
PC8253-25G
PC8253-5G
PC8253-1G
APOSPC8253-25GEA
306
EUR
APOSPC8253-25G
APOSPC8253-5G
APOSPC8253-1G
4-(Trifluoromethylthio)phenylacetic acid 96%
4-(Trifluoromethylthio)phenylacetic acid
Formel:
C₉H₇F₃O₂S Molecular Weight: 236.208 g/mol Schmelzpunkt: 114…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236351 CAS-Nummer: 243977-23-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...