1-Iodo-1H,1H,2H,2H-perfluoro(4,7,10,13,16-pentamethyl-5,8,11,14,17-pentaoxaeicosane) 95%
Lieferant: Apollo Scientific
PC8266-1G
PC8266-5G
APOSPC8266-1GEA
158
EUR
APOSPC8266-1G
APOSPC8266-5G
1-Iodo-1H,1H,2H,2H-perfluoro(4,7,10,13,16-pentamethyl-5,8,11,14,17-pentaoxaeicosane) 95%
1-Iodo-1H,1H,2H,2H-perfluoro(4,7,10,13,16-pentamethyl-5,8,11,14,17-pentaoxaeicosane)
Formel:
C₂₀H₄F₃₇IO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08460486 CAS-Nummer: 1212369-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...