Ethyl-3-(trifluoromethyl)-2-pyridinecarboxylate

Lieferant: Apollo Scientific
Achtung

PC8272-1G
APOSPC8272-1GEA 270 EUR
APOSPC8272-1G
Ethyl-3-(trifluoromethyl)-2-pyridinecarboxylate
Ethyl-3-(trifluoromethyl)-2-pyridinecarboxylate
Formel: C₉H₈F₃NO₂
Molecular Weight: 219,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864691
CAS-Nummer: 952182-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR