3-Brom-5-[(2-methyl-1H-imidazol-1-yl)sulfonyl]benzotrifluorid

Lieferant: Apollo Scientific

PC8277-1G
APOSPC8277-1GEA 357 EUR
APOSPC8277-1G
3-Brom-5-[(2-methyl-1H-imidazol-1-yl)sulfonyl]benzotrifluorid
3-Brom-5-[(2-methyl-1H-imidazol-1-yl)sulfonyl]benzotrifluorid
Formel: C₁₁H₈BrF₃N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09801016
CAS-Nummer: 951884-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR