1-((3-Brom-5-(trifluormethyl)phenyl)sulfonyl)piperidin
Lieferant: Apollo Scientific
Synonyme:
3-Bromo-5-(piperidin-1-ylsulphonyl)benzotrifluoride
PC8278-1G
APOSPC8278-1GEA
248
EUR
APOSPC8278-1G
1-((3-Brom-5-(trifluormethyl)phenyl)sulfonyl)piperidin
1-((3-Brom-5-(trifluormethyl)phenyl)sulfonyl)piperidin
Formel:
C₁₂H₁₃BrF₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09801013 CAS-Nummer: 951884-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...