1-(3-Brom-5-(trifluormethyl)phenylsulfonyl)pyrrolidin

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-5-(pyrrolidin-1-ylsulphonyl)benzotrifluoride

PC8279-1G
APOSPC8279-1GEA 248 EUR
APOSPC8279-1G
1-(3-Brom-5-(trifluormethyl)phenylsulfonyl)pyrrolidin
1-(3-Brom-5-(trifluormethyl)phenylsulfonyl)pyrrolidin
Formel: C₁₁H₁₁BrF₃NO₂S
Molecular Weight: 358,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801012
CAS-Nummer: 951884-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR