4-Amino-3,5-difluorphenol
Lieferant: Apollo Scientific
|
Achtung
|
PC8306-1G
APOSPC8306-1GEA
364
EUR
APOSPC8306-1G
4-Amino-3,5-difluorphenol
4-Amino-3,5-difluorphenol
Formel:
C₆H₅F₂NO Schmelzpunkt: 144…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094491 CAS-Nummer: 135086-76-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...