4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC8316-1G
APOSPC8316-1GEA
139
EUR
APOSPC8316-1G
4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide 97%
4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide
Formel:
C₈H₈F₃N₃S Schmelzpunkt: 120…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041291 CAS-Nummer: 20069-30-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...