2,6-Difluor-4-hydroxybenzonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyano-3,5-difluorophenol

PC8318-1G PC8318-25G PC8318-5G
APOSPC8318-1GEA 58.2 EUR
APOSPC8318-1G APOSPC8318-25G APOSPC8318-5G
2,6-Difluor-4-hydroxybenzonitril 98%
2,6-Difluor-4-hydroxybenzonitril
Formel: C₇H₃F₂NO
Schmelzpunkt: 125…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094499
CAS-Nummer: 123843-57-2
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR