4-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid

Lieferant: Apollo Scientific

Synonyme: 4-Boronophenyl 4-fluoro-2-nitrophenyl ether

PC8322-1G PC8322-5G
APOSPC8322-1GEA 244 EUR
APOSPC8322-1G APOSPC8322-5G
4-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid
4-(4-Fluoro-2-nitrophenoxy)benzeneboronic acid
Formel: C₁₂H₉BFNO₅
Molecular Weight: 277.013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800895
CAS-Nummer: 957062-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR