Terephthalbis(4-fluoroaniline) 98%

Lieferant: Apollo Scientific

Synonyme: 1,1'-(1,4-Phenylene)bis(N-(4-fluorophenyl)methanimine), Bis(4-fluoroanilino)terephthalaldehyde

PC8325-10G
APOSPC8325-10GEA 553 EUR
APOSPC8325-10G
Terephthalbis(4-fluoroaniline) 98%
Terephthalbis(4-fluoroaniline)
Formel: C₂₀H₁₄F₂N₂
Molecular Weight: 320.34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017952
CAS-Nummer: 17866-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR