5-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]-1,3,4-thiadiazol-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-(3,4,4-trifluorobut-3-en-1-ylthio)-1,3,4-thiadiazole
PC8328-1G
PC8328-250MG
PC8328-5G
APOSPC8328-1GEA
173
EUR
APOSPC8328-1G
APOSPC8328-250MG
APOSPC8328-5G
5-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]-1,3,4-thiadiazol-2-amine
5-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]-1,3,4-thiadiazol-2-amine
Formel:
C₆H₆F₃N₃S₂ Schmelzpunkt: 160…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01443829 CAS-Nummer: 145070-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...